FGR Recombinant Human Protein DFS Item
FGR Recombinant Human Protein
Manufacturer:  Life Technologies LSP3041
 View more versions of this product
Catalog No. P3041
Description
Description
FGR is a SRC-related kinase present in hematopoietic cells. FGR signals within multiple pathways including within the integrin pathway and downstream of chemokine and cytokine receptors.
Specifications
Specifications
NP_005239
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
62kDa
80% by SDS-PAGE
–80°C
1 Each
FGR
FGR was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.5), 10mM MgCl2, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
SRC
370nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL
His Tag
0.18mg/mL
2268
FGR Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Cytoplasmic tyrosine kinase
FGR Recombinant Human Protein
Kinase assay
Baculovirus
FGR Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.