Invitrogen™ FGR Recombinant Human Protein DFS Item
FGR is a SRC-related kinase present in hematopoietic cells.
Manufacturer:  Invitrogen™ LSPV4078
 View more versions of this product
Catalog No. PV4078
Description
Description
FGR signals within multiple pathways including within the integrin pathway and downstream of chemokine and cytokine receptors.
Specifications
Specifications
NP_005239
Frozen
62kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
SRC Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
20mM Tris, pH 7.5, 100mM NaCl, 0.05mM EDTA, 0.05% NP-40, 1mM DTT, 50% glycerol
1mg
RUO - research use only
FGR
Baculovirus
Human
His Tag
Documents