Please login to your online account to display your discounted pricing

FGIN-1-43, Tocris


Potent, specific ligand for mitochondrial DBI receptor

Quantity 10mg

Manufacturer: r & d systems  065910

 View more versions of this product

Catalog No. 065910

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 487.51
Assay Percent Range >98%
CAS 145040-29-5
Chemical Name or Material FGIN-1-43
Molecular Formula C28H36Cl2N2O
Synonym N, N-Dihexyl-2-(4-chlorophenyl)-5-chloroindole-3-acetamide
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material FGIN-1-43
Assay Percent Range >98%
Molecular Formula C28H36Cl2N2O
Formula Weight 487.51
Synonym N, N-Dihexyl-2-(4-chlorophenyl)-5-chloroindole-3-acetamide
Accessories

Store at Room Temperature