Please login to your online account to display your discounted pricing

FGIN-1-43, Tocris

Potent, specific ligand for mitochondrial DBI receptor

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material FGIN-1-43
Assay Percent Range >98%
Molecular Formula C28H36Cl2N2O
Formula Weight 487.51
Synonym N, N-Dihexyl-2-(4-chlorophenyl)-5-chloroindole-3-acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

065910

 
r & d systems
065910
10mg Each for $168.15

065950

 
r & d systems
065950
50mg Each for $697.40
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 487.51
Assay Percent Range >98%
CAS 145040-29-5
Chemical Name or Material FGIN-1-43
Molecular Formula C28H36Cl2N2O
Synonym N, N-Dihexyl-2-(4-chlorophenyl)-5-chloroindole-3-acetamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature