Please login to your online account to display your discounted pricing

FGIN-1-27, Tocris

Potent, specific ligand for mitochondrial DBI receptor

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material FGIN-1-27
Assay Percent Range >98%
Molecular Formula C28H37FN2O
Formula Weight 436.61
Synonym N, N-Dihexyl-2-(4-fluorophenyl)indole-3-acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

065810

 
r & d systems
065810
10mg Each for $168.15

065850

 
r & d systems
065850
50mg Each for $697.40
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 436.61
Assay Percent Range >98%
Chemical Name or Material FGIN-1-27
Molecular Formula C28H37FN2O
CAS 142720-24-9
Synonym N, N-Dihexyl-2-(4-fluorophenyl)indole-3-acetamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature