Please login to your online account to display your discounted pricing

FGIN-1-27, Tocris


Potent, specific ligand for mitochondrial DBI receptor

Quantity 50mg

Manufacturer: r & d systems  065850

 View more versions of this product

Catalog No. 065850

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material FGIN-1-27
Quantity 50mg
CAS 142720-24-9
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C28H37FN2O
Formula Weight 436.61
Synonym N, N-Dihexyl-2-(4-fluorophenyl)indole-3-acetamide

Specifications

Chemical Name or Material FGIN-1-27
Assay Percent Range >98%
Molecular Formula C28H37FN2O
Formula Weight 436.61
Synonym N, N-Dihexyl-2-(4-fluorophenyl)indole-3-acetamide
Accessories

Store at Room Temperature