Invitrogen™ FGFR3 Recombinant Human Protein DFS Item
FGFR3 is a member of the FGFR family of receptor tyrosine kinases.
Manufacturer:  Invitrogen™ LSPV4107
 View more versions of this product
Catalog No. PV4107
Description
Description
The FGFR family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy.
Specifications
Specifications
NP_000133
Frozen
49.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
FGF Receptor Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 100mM NaCl, 0.2mM EDTA, 0.05% Triton X-100, 5mM DTT, 50% glycerol
1mg
RUO - research use only
FGFR3, CEK2, JTK4
Baculovirus
Human
His Tag
Documents