FGFR2 Recombinant Human Protein DFS Item
FGFR2 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3368
 View more versions of this product
Catalog No. PV3368
Description
Description
FGFR2 is a member of the FGFR family of receptor tyrosine kinases. This family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy.
Specifications
Specifications
NP_075261
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
52.1kDa
85% by SDS-PAGE
–80°C
1 Each
FGFR2, CEK3, TY14, BEK
FGFR2 was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
FGF receptor
FGFR2 Recombinant Human Protein
Human
0.43mg/mL
2263
FGFR2 Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Receptor tyrosine kinases
FGFR2 Recombinant Human Protein
Kinase assay
Baculovirus
Amino acids 403-822
2,700nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1μg/mL
His Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.