Please login to your online account to display your discounted pricing

Fexofenadine hydrochloride, Tocris

H1 receptor antagonist; non-sedating antiallergic agent

$129.05 - $546.19

Chemical Identifiers

Chemical Name or Material Fexofenadine hydrochloride
Assay Percent Range >99%
Molecular Formula C32H39NO4·HCl
Formula Weight 538.12
Synonym α, α-Dimethyl-4-[1-hydroxy-4-[4-(hydroxydiphenylmethyl)-1-piperidinyl]butyl]-benzeneacetic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

242910

 
r & d systems
RDS242910
10mg Each for $129.05

242950

 
r & d systems
RDS242950
50mg Each for $546.19
Description & Specifications

Specifications

Formula Weight 538.12
Quantity 10mg
Chemical Name or Material Fexofenadine hydrochloride
Molecular Formula C32H39NO4·HCl
Synonym α, α-Dimethyl-4-[1-hydroxy-4-[4-(hydroxydiphenylmethyl)-1-piperidinyl]butyl]-benzeneacetic acid hydrochloride
Recommended Storage Desiccate at +4°C
Assay Percent Range >99%
CAS 153439-40-8
Safety and Handling

Desiccate at +4°C