Please login to your online account to display your discounted pricing

Alfa Aesar™ Fetuin I, Fetal Bovine Serum

For research use

Specifications

Formulation Liquid
Recombinant No
Storage Requirements 2° to 8°C
Source Human plasma
Product Type DiO-Lipoprotein, low density, Human Plasma
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64267AMH

 
alfa aesar™
J64267AMH
200μg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formulation Liquid
Recombinant No
Storage Requirements 2° to 8°C
Source Human plasma
Product Type DiO-Lipoprotein, low density, Human Plasma
Format Liquid
Regulatory Status RUO