Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ α-Fetoprotein (AFP) Ab-2, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ α-Fetoprotein (AFP) Ab-2, Rabbit Polyclonal Antibody.

$323.96 - $485.08

Specifications

Antigen alpha Fetoprotein (AFP) Ab-2
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

RB365A

 
thermo scientific™ lab vision™
RB365A
1mL Each for $485.08

RB365A1

 
thermo scientific™ lab vision™
RB365A1
500µL Each for $323.96
Description & Specifications

Specifications

Antigen alpha Fetoprotein (AFP) Ab-2
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rabbit
Host Species Rabbit
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Isotype IgG

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ α-Fetoprotein (AFP) Ab-2, Rabbit Polyclonal Antibody.

AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Pig and Sheep. Does not react with dog. Others not known.

Epitope: Not determined

Immunogen: Purified α-fetoprotein

Molecular Weight: 70 kDa

Positive Control: HEP-G-2 cells, Fetal liver or hepatocellular CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO