Gibco™ Fetal Bovine Serum, Qualified One Shot™, Australia origin DFS Item
Introducing the new, aliquot-free Gibco™ One Shot™ FBS 50mL bottle. Designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it. And that improves your chances for consistent results.
$161.00 - $1,512.00
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A3161001 Gibco™
LSA3161001
50mL Each for $161.00
A3161002 Gibco™
LSA3161002
10 x 50mL Each for $1,512.00
Specifications
Description
If you’re one of the more than 80% of researchers who aliquots their FBS into 50mL conical tubes, your life is about to change for the better. Transferring your FBS takes away your control — of how you spend your time, your contamination risk, and your volume consistency. It also wastes costly material.
The new Gibco™ One Shot™ FBS 50mL bottle design eliminates the need to aliquot, and the variability that comes with it. And that improves your chances for consistent results.
Product Features:
  • Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
  • Engineered to freeze/thaw without splitting or breaking
  • Fits in test tube racks and frees up freezer space
  • No labeling mishaps with our easy-to-read package label has all the information you need
  • Generates 33% less waste than aliquoting from 500mL bottles