FES (FPS) Recombinant Human Protein DFS Item
FES (FPS) Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3354
Catalog No. PV3354
Description
Description
FES (FPS) is a tyrosine protein kinase that is required for maintenance of cellular transformation. Its chromosomal location is linked to a specific translocation event identified in patients with acute promyelocytic leukemia, and it is also involved in normal hematopoiesis.
Specifications
Specifications
NP_001996
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
98.3kDa
85% by SDS-PAGE
–80°C
1 Each
FES (FPS)
FES was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
FER
1580nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1μg/mL
His Tag
0.27mg/mL
2242
FES (FPS) Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Cytoplasmic tyrosine kinase
FES (FPS) Recombinant Human Protein
Kinase assay
Baculovirus
FES (FPS) Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.