missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate Heptahydrate, ACS Reagent (Iron (II) Sulfate Heptahydrate), Reagents

Ferrous Sulfate Heptahydrate, ACS Reagent (Iron (II) Sulfate Heptahydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

$87.97 - $3848.27

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym Iron(II) Sulfate Heptahydrate
IUPAC Name λ²-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-337-961
View Documents
Labchem Inc
C1207000100C2
100 g
Each for $87.97
Only null left
 
01-337-960
View Documents
Labchem Inc
C120700010KG
10 kg
Each for $2,164.63
Only null left
 
01-337-959
View Documents
Labchem Inc
C120700025A5
2.5 kg
Each for $673.57
Only null left
 
01-337-958
View Documents
Labchem Inc
C120700025F4
25 kg
Each for $3,848.27
Only null left
 
01-337-957
View Documents
Labchem Inc
C1207000500A5
500 g
Each for $108.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7782-63-0
278.01
SURQXAFEQWPFPV-UHFFFAOYSA-L
λ²-iron(2+) heptahydrate sulfate
FeH14O11S
MFCD00149719
Iron(II) Sulfate Heptahydrate
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

>300°C
7782-63-0
100 g
MFCD00149719
SURQXAFEQWPFPV-UHFFFAOYSA-L
λ²-iron(2+) heptahydrate sulfate
ACS
1.897
Green or Bluish-Green
Solid
FeH14O11S
Iron(II) Sulfate Heptahydrate
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
278.01
Glass Bottle
Ferrous Sulfate Heptahydrate, ACS Reagent (Iron (II) Sulfate Heptahydrate)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'