Please login to your online account to display your discounted pricing

Ferrous Sulfate ACS AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Ferrous Sulfate
Name Note 7-Hydrate
CAS 7782-63-0
Purity Grade ACS Grade
Physical Form Granular
Molecular Formula FeSO4·7H2O
Formula Weight 278.02
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK5056212

 
avantor performance
505606
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK5056500

 
avantor performance
505612
POLYSTORMOR 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 278.02
Purity Grade ACS Grade
Quantity 2.5kg
Physical Form Granular
Name Note 7-Hydrate
CAS 7782-63-0
Packaging Poly
Chemical Name or Material Ferrous Sulfate
Molecular Formula FeSO4·7H2O