Please login to your online account to display your discounted pricing

Ferrous Sulfate, 7-Hydrate, Crystal, U.S.P. - F.C.C., J.T.Baker™

$204.61 - $558.10

Chemical Identifiers

Chemical Name or Material Ferrous Sulfate
CAS 7782-63-0
Name Note 7-Hydrate
Purity Grade FCC/USP
Molecular Formula FeSO4·7H2O
Formula Weight 278.02
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002877

 
avantor performance
207405
Wide Mouth Amber Glass Bottle 2.5kg Each for $558.10

02002878

 
avantor performance
207401
Wide Mouth Amber Glass Bottle 500g Each for $204.61
Description & Specifications

Specifications

Purity Grade FCC/USP
Physical Form Crystals
Formula Weight 278.02
CAS 7782-63-0
Molecular Formula FeSO4·7H2O
Name Note 7-Hydrate
Quantity 2.5kg
Specific Gravity 1L = 1.90kg
Chemical Name or Material Ferrous Sulfate
Packaging Wide Mouth Amber Glass Bottle