Please login to your online account to display your discounted pricing

Ferrous Sulfate, 7-Hydrate, Crystal, Multi-Compendial, J.T.Baker™

$188.10 - $188.10

Chemical Identifiers

Chemical Name or Material Ferrous Sulfate
CAS 7782-63-0
Name Note 7-Hydrate
Purity Grade Multi-Compendial
Molecular Formula FeSO4·7H2O
Formula Weight 278.02
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002879

 
avantor performance
206301
Wide Mouth Amber Glass Bottle 500g Each for $188.10
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 278.02
Chemical Name or Material Ferrous Sulfate
CAS 7782-63-0
Molecular Formula FeSO4·7H2O
Quantity 500g
Specific Gravity 1L = 1.90kg
Purity Grade Multi-Compendial
Packaging Wide Mouth Amber Glass Bottle
Name Note 7-Hydrate