missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ferrous Ammonium Sulfate Solution, Certified, 0.497N ±0.010N, LabChem™

$136.74 - $136.74

Chemical Identifiers

CAS 7732-18-5,7783-85-9,7664-93-9
CAS Max % 79.5,19.5,1
Linear Formula Fe(NH4)2(SO4)2·6H2O
Formula Weight 392.14
UN Number UN3264
Synonym FAS
Chemical Name or Material Ferrous Ammonium Sulfate Solution
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC145452 Lab Chem
LC145452
1L Each for $136.74
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7783-85-9,7664-93-9
Fe(NH4)2(SO4)2·6H2O
UN3264
Ferrous Ammonium Sulfate Solution
79.5,19.5,1
392.14
FAS
Certified Grade

Specifications

392.14
Room Temperature
Gaseous Ammonia; Sulfur compounds; Thermal decomposition generates: Corrosive vapors
0.497N ±0.010N
Liquid
FAS
Fe(NH4)2(SO4)2·6H2O
UN3264
Certified Grade
7732-18-5,7783-85-9,7664-93-9
Clear, light green
1L
Amber Glass
79.5,19.5,1
Soluble in water
Passes Test
Ferrous Ammonium Sulfate Solution

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger