Please login to your online account to display your discounted pricing

Ferrous Ammonium Sulfate, Hexahydrate, ACS Grade, 98.5% to 101.5%, LabChem™

$222.86 - $222.86

Chemical Identifiers

Chemical Name or Material Ferrous Ammonium Sulfate
CAS 7783-85-9
CAS Max % 100
Name Note Hexahydrate
Purity Grade ACS Grade
Linear Formula Fe(NH4)2(SO4)2·6H2O
Formula Weight 392.14
Synonym FAS
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC144601

 
lab chem
LC144601
500g Each for $222.86
Description & Specifications

Specifications

Assay 98.5 to 101.5%
Formula Weight 392.14
Chemical Name or Material Ferrous Ammonium Sulfate
Identification Passes Test
Name Note Hexahydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Solubility Soluble in water
Specific Gravity 1.865g/cm3
Storage Information Room Temperature
Linear Formula Fe(NH4)2(SO4)2·6H2O
CAS 7783-85-9
Decomposition Information Gaseous Ammonia; Sulfur compounds
Synonym FAS
Color Pale green
Melting Point 100°C
Quantity 500g
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Density 1.865g/cm3
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.