Please login to your online account to display your discounted pricing

Ferrous Ammonium Sulfate, For Nitrogen (Nitrite), Certified, 0.0500M ±0.0005M (0.05N), LabChem™

$111.74 - $111.74

Chemical Identifiers

Chemical Name or Material Ferrous Ammonium Sulfate
CAS 7732-18-5
CAS Max % 96.04
CAS 7664-93-9
CAS Max % 2
CAS 7783-85-9
CAS Max % 1.96
Name Note For Nitrogen (Nitrite)
Purity Grade Certified Grade
Linear Formula Fe(NH4)2(SO4)2·6H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC144802

 
lab chem
LC144802
1L Each for $111.74
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 392.14
Chemical Name or Material Ferrous Ammonium Sulfate
Traceability to NIST Traceable to NIST
CAS Max % 1.96
Decomposition Information Gaseous Ammonia; Sulfur compounds; Thermal decomposition generates: Corrosive vapors
Color pale green
Quantity 1L
Name Note For Nitrogen (Nitrite)
Physical Form Liquid
Concentration 0.0500M ±0.0005M (0.05N)
Linear Formula Fe(NH4)2(SO4)2·6H2O
CAS 7732-18-5
UN Number UN3264
Synonym FAS
Health Hazard 1 Danger
CAS Max % 96.04
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Identification Passes Test
CAS 7664-93-9
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Amber Glass
CAS Max % 2
Solubility Soluble in water
CAS 7783-85-9
Storage Information Room Temperature
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.