Please login to your online account to display your discounted pricing

Ferrous Ammonium Sulfate, Certified, 0.40N ±0.01N (0.4M), LabChem™

$308.63 - $308.63

Chemical Identifiers

Chemical Name or Material Ferrous Ammonium Sulfate
CAS 7732-18-5
CAS Max % 81.3
CAS 7783-85-9
CAS Max % 15.7
CAS 7664-93-9
CAS Max % 3
Purity Grade Certified Grade
Linear Formula Fe(NH4)2(SO4)2·6H2O
Formula Weight 392.14
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC145404

 
lab chem
LC145404
4L Each for $308.63
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 392.14
CAS 7664-93-9
Physical Form Liquid
Packaging Amber Glass
Identification Passes Test
Storage Information Room Temperature
Chemical Name or Material Ferrous Ammonium Sulfate
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Color Clear pale green
Synonym FAS
Concentration 0.40N ±0.01N (0.4M)
Linear Formula Fe(NH4)2(SO4)2·6H2O
Health Hazard 1 Danger
Quantity 4L
UN Number UN3264
CAS Max % 3
CAS 7783-85-9
Solubility Soluble in water
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
CAS Max % 15.7
CAS 7732-18-5
CAS Max % 81.3
Decomposition Information Gaseous Ammonia; Sulfur compounds; Thermal decomposition generates: Corrosive vapors
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.