Please login to your online account to display your discounted pricing

Ferrous Ammonium Sulfate, 6-Hydrate, Fine Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 1kg
Packaging Poly Bottle

Manufacturer: avantor performance  205419

 View more versions of this product

Catalog No. 02002882

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 392.13
Chemical Name or Material Ferrous Ammonium Sulfate
Name Note 6-Hydrate
Physical Form Fine Crystal
Molecular Formula Fe(NH4)2(SO4)2·6H2O
CAS 7783-85-9
Quantity 1kg
Packaging Poly Bottle

Specifications

Chemical Name or Material Ferrous Ammonium Sulfate
CAS 7783-85-9
Name Note 6-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Fe(NH4)2(SO4)2·6H2O
Formula Weight 392.13