missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ferrous Ammonium Sulfate, 0.00282 Normal (0.00282 Molar), 1mL = 0.1mg Cl, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI314432

 View more versions of this product

Catalog No. 3144-32



Description

Description

Specifications

Specifications

Ferrous Ammonium Sulfate
7783-85-9
Liquid
Green
0.00282N,0.00282M
1 mL = 0.1mg Cl2
Amber Glass
1L

Chemical Identifiers

7783-85-9
SDS Documents