Please login to your online account to display your discounted pricing

Alfa Aesar™ (Ferrocenylmethyl)trimethylammonium iodide

$807.62 - $807.62

Chemical Identifiers

Chemical Name or Material (Ferrocenylmethyl)trimethylammonium iodide
CAS 12086-40-7
Formula Weight 385.08
MDL Number MFCD00041534
Synonym N,N-Dimethylaminomethylferrocene methiodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3939906

  SDS
alfa aesar™
3939906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3939914

  SDS
alfa aesar™
3939914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3939922

  SDS
alfa aesar™
3939922
100g Each for $807.62
Description & Specifications

Specifications

Formula Weight 385.08
Chemical Name or Material (Ferrocenylmethyl)trimethylammonium iodide
Physical Form Crystalline
Quantity 5g
Melting Point 220°C (dec.)
CAS 12086-40-7
MDL Number MFCD00041534
Synonym N,N-Dimethylaminomethylferrocene methiodide