Please login to your online account to display your discounted pricing

Alfa Aesar™ Ferrocenecarboxylic acid, 98%

$183.96 - $659.26

Chemical Identifiers

Chemical Name or Material Ferrocenecarboxylic acid
CAS 1271-42-7
Molecular Formula C11H10FeO2
Formula Weight 230.05g/mol
MDL Number MFCD00001430
Synonym Carboxyferrocene; (Carboxycyclopentadienyl)cyclopentadienyliron
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1687406

 
alfa aesar™
A16874-06
5g Each for $183.96

AAA1687414

 
alfa aesar™
A16874-14
25g Each for $659.26
Description & Specifications

Specifications

Melting Point ≈210°C (dec.)
Quantity 5g
Chemical Name or Material Ferrocenecarboxylic acid
CAS 1271-42-7
Assay 98%
Molecular Formula C11H10FeO2
Formula Weight 230.05g/mol
MDL Number MFCD00001430
Synonym Carboxyferrocene; (Carboxycyclopentadienyl)cyclopentadienyliron
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
Ferrocenecarboxylic acid is used to esterify complex mixtures of phenols and alcohols for analysis by GCMS. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.