Please login to your online account to display your discounted pricing

Alfa Aesar™ Ferrocenecarboxaldehyde, 97%

Chemical Identifiers

Chemical Name or Material Ferrocenecarboxaldehyde
CAS 12093-10-6
Formula Weight 214.05
MDL Number MFCD00001429
Synonym [Cyclopentadienyl(formylcyclopentadienyl)iron]
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3925409

  SDS
alfa aesar™
3925409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 214.05
Assay 97%
MDL Number MFCD00001429
Chemical Name or Material Ferrocenecarboxaldehyde
TSCA TSCA
Quantity 10g
Physical Form Crystalline
Synonym [Cyclopentadienyl(formylcyclopentadienyl)iron]
Sensitivity Air Sensitive
CAS 12093-10-6
Melting Point 124°C