Learn More
Ferroceneacetonitrile, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 4413103
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ferroceneacetonitrile | |
82°C | |
MFCD00068572 | |
UN3439 | |
Ferroceneacetonitrile | |
225.08 | |
Powder |
1316-91-2 | |
C12H11FeN | |
1 g | |
[Fe].c1cccc1.N#CCc1cccc1 | |
225.07 | |
97% |
Chemical Identifiers
1316-91-2 | |
225.07 | |
Ferroceneacetonitrile |
C12H11FeN | |
MFCD00068572 | |
[Fe].c1cccc1.N#CCc1cccc1 |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures