missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ferrocene, 99%

Manufacturer:  Alfa Aesar™ 87202A1

 View more versions of this product

Catalog No. AA87202A1



Description

Description

  • Packaged under Argon
Specifications

Specifications

Ferrocene
Air & Moisture Sensitive
Yellow to Orange
1kg
C10H10Fe-2
UN1325
ferrocene, bis cyclopentadienyl iron
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Fe]
186.035
186.04
Characteristic
249°C
173°C to 174°C
102-54-5
MFCD00001427
14,4037
UEWKINKPISJKRD-UHFFFAOYSA-N
cyclopenta-1,3-diene;iron
25199998
99%

Chemical Identifiers

102-54-5
186.035
UEWKINKPISJKRD-UHFFFAOYSA-N
25199998
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Fe]
C10H10Fe-2
MFCD00001427
ferrocene, bis cyclopentadienyl iron
cyclopenta-1,3-diene;iron
Safety and Handling

Safety and Handling

GHS H Statement
H228-H302
Flammable solid.
Harmful if swallowed.

GHS P Statement
P210-P280-P240-P241-P301+P312-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : LK0700000

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

SDS Documents