Please login to your online account to display your discounted pricing

Ferrocene, 98%, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC119140025

 View more versions of this product

Catalog No. AC119140025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder or Crystals
Assay ≥97.5%
Formula Weight 186.04
Quantity 2.5kg
Color Orange-brown
Melting Point 173° to 176°C
Sublimation Point 0100.0°C
Chemical Name or Material Ferrocene
Infrared Spectrum Authentic
Packaging Plastic bottle
Additional Information product may become lumpy during storage
Synonym Dicyclopentadienyliron
CAS 102-54-5
Molecular Formula C10H10Fe
Assay Percent Range 98%
Health Hazard 1 Warning

Specifications

Chemical Name or Material Ferrocene
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H10Fe
Formula Weight 186.04
Synonym Dicyclopentadienyliron
Accessories

Warning

SDS & Safety Documents