Please login to your online account to display your discounted pricing

Alfa Aesar™ Ferriprotoporphyrin IX chloride

Chemical Identifiers

Chemical Name or Material Ferriprotoporphyrin IX chloride
CAS 15489-47-1
Formula Weight 651.93g/mol
MDL Number MFCD00010726
Synonym Hemin chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3206103

 
alfa aesar™
3206103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3206106

 
alfa aesar™
32061-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Quantity 1g
Chemical Name or Material Ferriprotoporphyrin IX chloride
CAS 15489-47-1
Physical Form Crystalline
Formula Weight 651.93g/mol
MDL Number MFCD00010726
Synonym Hemin chloride