Please login to your online account to display your discounted pricing

Ferric Sulfate, n-Hydrate, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$1,333.32 - $1,333.32

Chemical Identifiers

Chemical Name or Material Ferric Sulfate
CAS 15244-10-7
Name Note n-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Fe2(SO4)3·nH2O
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002869

 
avantor performance
204605
Poly Bottle 2.5kg Each for $1,333.32
Description & Specifications

Specifications

Chemical Name or Material Ferric Sulfate
CAS 15244-10-7
Packaging Poly Bottle
Molecular Formula Fe2(SO4)3·nH2O
Name Note n-Hydrate
Specific Gravity 1L = 3.10kg
Quantity 2.5kg
Purity Grade BAKER ANALYZED™ Reagent