missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ferric Sulfate, MP Biomedicals
Manufacturer: Mp Biomedicals 0215804283
Specifications
Ferric Sulfate | |
Fe2O12S3 | |
RUTXIHLAWFEWGM-UHFFFAOYSA-H | |
iron(3+);trisulfate | |
24826 | |
399.9 |
10028-22-5 | |
diiron tris sulphate, diiron trisulfate, ferric persulfate, ferric sesquisulfate, ferric sulfate, ferric tersulfate, iron iii sulfate, iron persulfate, iron sesquisulfate, iron tersulfate | |
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3] | |
399.858 | |
CHEBI:53438 | |
250g |
Chemical Identifiers
10028-22-5 | |
399.858 | |
diiron tris sulphate, diiron trisulfate, ferric persulfate, ferric sesquisulfate, ferric sulfate, ferric tersulfate, iron iii sulfate, iron persulfate, iron sesquisulfate, iron tersulfate | |
CHEBI:53438 | |
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3] |
Fe2O12S3 | |
RUTXIHLAWFEWGM-UHFFFAOYSA-H | |
24826 | |
iron(3+);trisulfate |
Safety and Handling
Recommended Storage : Room Temperature