Please login to your online account to display your discounted pricing

Ferric Oxide, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  202405

 View more versions of this product

Catalog No. 02002867

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 159.69
CAS 1309-37-1
Molecular Formula Fe2O3
Physical Form Powder
Quantity 2.5kg
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 5.24kg
Packaging Poly Bottle
Chemical Name or Material Ferric Oxide

Specifications

Chemical Name or Material Ferric Oxide
CAS 1309-37-1
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Fe2O3
Formula Weight 159.69