Please login to your online account to display your discounted pricing

Ferric Oxide, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$189.74 - $488.45

Chemical Identifiers

Chemical Name or Material Ferric Oxide
CAS 1309-37-1
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Fe2O3
Formula Weight 159.69
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002867

 
avantor performance
202405
Poly Bottle 2.5kg Each for $488.45

02002868

 
avantor performance
202401
Wide Mouth Amber Glass Bottle 500g Each for $189.74
Description & Specifications

Specifications

Formula Weight 159.69
CAS 1309-37-1
Molecular Formula Fe2O3
Physical Form Powder
Quantity 2.5kg
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 5.24kg
Packaging Poly Bottle
Chemical Name or Material Ferric Oxide