missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ferric Nitrate Nonahydrate, (Crystalline),MP Biomedicals

$99.14 - $99.14

Chemical Identifiers

CAS 7782-61-8
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.992
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, ferric nitrate, nonahydrate, ferricnitratenonahydrate, iron iii nitrate, iron iii nitrate hydrate, iron iii nitrate nonahydrate, iron, reference standard solution, nitric acid, iron 3+ salt, nonahydrate, unii-vmx4uop3vn, vmx4uop3vn
PubChem CID 16211566
IUPAC Name iron(3+);trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15804190
View Documents
Mp Biomedicals
0215804190
500g
Each for $99.14
 
Description

Description

Specifications

Chemical Identifiers

7782-61-8
403.992
ferric nitrate nonahydrate, ferric nitrate, nonahydrate, ferricnitratenonahydrate, iron iii nitrate, iron iii nitrate hydrate, iron iii nitrate nonahydrate, iron, reference standard solution, nitric acid, iron 3+ salt, nonahydrate, unii-vmx4uop3vn, vmx4uop3vn
iron(3+);trinitrate;nonahydrate
FeH18N3O18
SZQUEWJRBJDHSM-UHFFFAOYSA-N
16211566
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3]

Specifications

7782-61-8
ferric nitrate nonahydrate, ferric nitrate, nonahydrate, ferricnitratenonahydrate, iron iii nitrate, iron iii nitrate hydrate, iron iii nitrate nonahydrate, iron, reference standard solution, nitric acid, iron 3+ salt, nonahydrate, unii-vmx4uop3vn, vmx4uop3vn
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3]
403.992
404
500g
FeH18N3O18
SZQUEWJRBJDHSM-UHFFFAOYSA-N
iron(3+);trinitrate;nonahydrate
16211566
Crystalline
Ferric Nitrate Nonahydrate
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Product Certifications