missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ferric Nitrate, Nonahydrate, ACS Grade, 98.0% to 101.0%, LabChem™

$152.89 - $152.89

Chemical Identifiers

CAS 7782-61-8
CAS Max % 100
Linear Formula Fe(NO3)3·9H2O
Formula Weight 404.02
UN Number UN1466
Chemical Name or Material Ferric Nitrate, Nonahydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC144101 Lab Chem
LC144101
500g Each for $152.89
Specifications Description & Specifications

Chemical Identifiers

7782-61-8
Fe(NO3)3·9H2O
UN1466
ACS Grade
100
404.02
Ferric Nitrate, Nonahydrate

Specifications

7782-61-8
Crystals
404.02
Passes Test
Soluble in water
Nitrogen oxides
47°C
Ferric Nitrate, Nonahydrate
ACS Grade
100
Fe(NO3)3·9H2O
UN1466
Poly Bottle
Room Temperature
Pale violet
500g
98.0 to 101.0%

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Avoid breathing dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

Warning