Please login to your online account to display your discounted pricing

Ferric Nitrate, 9-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  201801

 View more versions of this product

Catalog No. 02002865

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 404
Physical Form Crystals
Purity Grade Baker Analyzed™ ACS Reagent
CAS 7782-61-8
Chemical Name or Material Ferric Nitrate
Packaging Poly Bottle
Quantity 500g
Molecular Formula Fe(NO3)3·9H2O
Name Note 9-Hydrate
Specific Gravity 1L = 1.68kg

Specifications

Chemical Name or Material Ferric Nitrate
CAS 7782-61-8
Name Note 9-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Fe(NO3)3·9H2O
Formula Weight 404