Please login to your online account to display your discounted pricing

Ferric Chloride, T.S., BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$34.45 - $34.45

Chemical Identifiers

Chemical Name or Material Ferric Chloride
Name Note T.S.
Purity Grade BAKER ANALYZED™ Reagent
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002861

 
avantor performance
592104
Narrow Mouth Amber Glass Bottle 100mL Each for $34.45
Description & Specifications

Specifications

Chemical Name or Material Ferric Chloride
Name Note T.S.
Packaging Narrow Mouth Amber Glass Bottle
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 1.00kg
Quantity 100mL