missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ferric Chloride Solution, For Sulfide, 100g in 40mL Water, LabChem™

$63.14 - $63.14

Chemical Identifiers

CAS 10025-77-1,7732-18-5
CAS Max % 71,29
Linear Formula FeCl3·6H2O
Formula Weight 270.3
UN Number UN2582
Chemical Name or Material Ferric Chloride Solution, For Sulfide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC143807 Lab Chem
LC143807
125mL Each for $63.14
Specifications Description & Specifications

Chemical Identifiers

10025-77-1,7732-18-5
FeCl3·6H2O
UN2582
71,29
270.3
Ferric Chloride Solution, For Sulfide

Specifications

270.3
100g in 40mL Water
Soluble in water
Passes Test
UN2582
Poly Bottle
Dark amber
Ferric Chloride Solution, For Sulfide
125mL
Liquid
Hydrogen chloride; Iron oxide; Thermal decomposition generates: Corrosive vapors
71,29
FeCl3·6H2O
10025-77-1,7732-18-5
Room Temperature

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life.

GHS P Statement
Do not breathe vapors, mist, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or air): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger