Please login to your online account to display your discounted pricing

Ferric Chloride Solution , For Sulfide, 100g in 40mL Water, LabChem™

$189.94 - $189.94

Chemical Identifiers

Chemical Name or Material Ferric Chloride Solution
CAS 10025-77-1
CAS Max % 71
CAS 7732-18-5
CAS Max % 29
Name Note For Sulfide
Linear Formula FeCl3·6H2O
Formula Weight 270.3
UN Number UN2582
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC143801

 
lab chem
LC143801
500mL Each for $189.94
Description & Specifications

Specifications

Formula Weight 270.3
Chemical Name or Material Ferric Chloride Solution
Decomposition Information Hydrogen chloride; Iron oxide; Thermal decomposition generates: Corrosive vapors
Color Dark amber
Quantity 500mL
Name Note For Sulfide
Physical Form Liquid
Concentration 100g in 40mL Water
Linear Formula FeCl3·6H2O
CAS 10025-77-1
UN Number UN2582
Health Hazard 1 Danger
CAS Max % 71
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life.
Identification Passes Test
CAS 7732-18-5
Health Hazard 3 GHS P Statement
Do not breathe vapors, mist, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or air): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
CAS Max % 29
Solubility Soluble in water
Storage Information Room Temperature
Safety and Handling

Danger

GHS P Statement
Do not breathe vapors, mist, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or air): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life.