Please login to your online account to display your discounted pricing

Ferric Chloride Solution, Fisher Chemical


Quantity 500mL
Packaging Amber Glass

Manufacturer: ricca chemical company  RISF9000500

 View more versions of this product

Catalog No. SF9000-500

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Methods For AOAC Method 937.05
Chemical Name or Material Ferric Chloride Solution
CAS Max % 0.25
CAS 7732-18-5
ChemAlert Storage Symbol White
CAS Min % 97.0
CAS Max % 100.0
CAS 10025-77-1
Packaging Amber Glass
CAS Min % 0.97
CAS Max % 1.01
CAS 7647-01-0
CAS Min % 0.24

Specifications

Chemical Name or Material Ferric Chloride Solution
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 10025-77-1
CAS Min % 0.97
CAS Max % 1.01
CAS 7647-01-0
CAS Min % 0.24
CAS Max % 0.25
Accessories