missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ferric Chloride Solution, 40% w/v (Laboratory), Fisher Chemical

Manufacturer:  Fisher Chemical SI1151

Catalog No. SI115-1




Description

Description

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Specifications

Specifications

Ferric Chloride Solution, 40% w/v
7705-08-0,7732-18-5
MFCD00011005
Liquid
Laboratory
3.85 to 6.6g/100mL
Glass Bottle
Brown
40% w/v
40%
Iron (III) Chloride Solution, 40% w/v
37.2 to 42.7g/100mL
37.2 to 42.7g/100mL
Pass test
1.45g/mol
1L

Chemical Identifiers

7705-08-0
MFCD00011005
Iron (III) Chloride Solution, 40% w/v
Safety and Handling

Safety and Handling

SDS Documents
Product Certifications