Please login to your online account to display your discounted pricing

Ferric Chloride Oxidizing Solution, For Cyanide, Certified, LabChem™

$60.71 - $60.71

Chemical Identifiers

Chemical Name or Material Ferric Chloride Oxidizing Solution
CAS 7732-18-5
CAS Max % 97.4
CAS 5329-14-6
CAS Max % 1.6
CAS 10025-77-1
CAS Max % 1
Name Note For Cyanide
Purity Grade Certified Grade
Linear Formula FeCl3·6H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC143301

 
lab chem
LC143301
500mL Each for $60.71
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 270.3
CAS 5329-14-6
Storage Information Room Temperature
Identification Passes Test
Decomposition Information Hydrogen chloride; Iron oxide; Sulfur compounds
Name Note For Cyanide
Solubility Soluble in water
Packaging Poly Bottle
CAS Max % 1.6
CAS 10025-77-1
Quantity 500mL
CAS Max % 1
CAS 7732-18-5
Chemical Name or Material Ferric Chloride Oxidizing Solution
Sensitivity Passes Test
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 97.4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life.
Physical Form Liquid
Color Clear yellow to light amber
Linear Formula FeCl3·6H2O
Safety and Handling

Warning

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life.