Please login to your online account to display your discounted pricing

Ferric Chloride Hexahydrate (Lumps/Technical), Fisher Chemical


Quantity 10kg
Packaging Poly Pail

Manufacturer: fisher scientific company  I8610

 View more versions of this product

Catalog No. I86-10

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 270.3
Boiling Point 280°C
Melting Point 37°C
Quantity 10kg
DOT Information DOT Class 8, : Corrosive
Physical Form Solid
Molecular Formula Cl3Fe · 6 H2O
Viscosity [NULL]
Color Dark Yellow
pH 2
Identification Pass test
Packaging Poly Pail
ChemAlert Storage Symbol White
Synonym Iron (III) Chloride
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Causes burns by all exposure routes. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:1
MDL Number MFCD00149712
Additional Information Vapor Pressure: Negligible
Chemical Name or Material Ferric Chloride Hexahydrate
CAS 10025-77-1

Specifications

Chemical Name or Material Ferric Chloride Hexahydrate
CAS 10025-77-1
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula Cl3Fe · 6 H2O
Formula Weight 270.3
MDL Number MFCD00149712
Synonym Iron (III) Chloride
Accessories
 

Emergency Overview
Causes burns by all exposure routes. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:1

DANGER!

SDS & Safety Documents