Please login to your online account to display your discounted pricing

Ferric Chloride Hexahydrate, ACS, LabChem™

$96.94 - $96.94

Chemical Identifiers

Chemical Name or Material Ferric Chloride Hexahydrate
CAS 10025-77-1
CAS Max % 100
Purity Grade ACS Grade
Linear Formula FeCl3·6H2O
Formula Weight 270.3
UN Number 3260
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC142901

 
lab chem
LC142901
500g Each for $96.94
Description & Specifications

Specifications

Assay 97.0 to 102.0%
Formula Weight 270.3
UN Number 3260
Linear Formula FeCl3·6H2O
CAS 10025-77-1
Purity Grade ACS Grade
Physical Form Solid
Color yellow-brown lumps
CAS Max % 100
Melting Point 37°C
Decomposition Information Hydrogen chloride; iron oxide
Quantity 500g
Packaging Poly Bottle
Storage Information Room Temperature
Chemical Name or Material Ferric Chloride Hexahydrate
Boiling Point 280°C
Identification Passes Test
Solubility Soluble in water