Please login to your online account to display your discounted pricing

Ferric Chloride, as Hexahydrate, Certified, 10.0% (w/v) ±0.5%, LabChem™

$40.71 - $40.71

Chemical Identifiers

Chemical Name or Material Ferric Chloride
CAS 7732-18-5
CAS Max % 75
CAS 7647-01-0
CAS Max % 15
CAS 10025-77-1
CAS Max % 10
Name Note as Hexahydrate
Purity Grade Certified Grade
Linear Formula FeCl3·6H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC143601

 
lab chem
LC143601
500mL Each for $40.71
Description & Specifications

Specifications

Purity Grade Certified Grade
Assay 10.0% ± 0.5% (w/v)
Formula Weight 270.3
Packaging Poly Bottle
CAS 7732-18-5
CAS 10025-77-1
CAS 7647-01-0
Physical Form Liquid
Quantity 500mL
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.
Decomposition Information Hydrogen chloride; Iron oxide; Thermal decomposition generates: Corrosive vapors
Storage Information Room Temperature
Identification Passes Test
Health Hazard 1 Danger
CAS Max % 15
Chemical Name or Material Ferric Chloride
CAS Max % 10
Linear Formula FeCl3·6H2O
Color Dark amber
UN Number UN3264
Name Note as Hexahydrate
CAS Max % 75
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.