missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ferric Chloride ACS AR Lumps, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 10025-77-1
Physical Form Lumps
Molecular Formula FeCl3·6H2O
Formula Weight 270.3
Chemical Name or Material Ferric Chloride, 6-Hydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK5029212 Avantor Performance
502906
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK5029500 Avantor Performance
502904*
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

10025-77-1
FeCl3·6H2O
Ferric Chloride, 6-Hydrate
Lumps
270.3
ACS Grade

Specifications

10025-77-1
FeCl3·6H2O
Poly
Ferric Chloride, 6-Hydrate
Lumps
270.3
2.5kg
ACS Grade