Please login to your online account to display your discounted pricing

Ferric Chloride ACS AR Lumps, Macron Fine Chemicals™


macron

Quantity 500g
Packaging POLYSTORMOR

Manufacturer: avantor performance  502904*

 View more versions of this product

Catalog No. MK5029500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 270.3
Purity Grade ACS Grade
Quantity 500g
Chemical Name or Material Ferric Chloride
Physical Form Lumps
CAS 10025-77-1
Molecular Formula FeCl3·6H2O
Name Note 6-Hydrate
Packaging POLYSTORMOR

Specifications

Chemical Name or Material Ferric Chloride
Name Note 6-Hydrate
CAS 10025-77-1
Purity Grade ACS Grade
Physical Form Lumps
Molecular Formula FeCl3·6H2O
Formula Weight 270.3