missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ferric Chloride, 10% (w/v) Aqueous Solution, Ricca Chemical
Manufacturer: Ricca Chemical Company RI31201
Specifications
Ferric Chloride Solution | |
10025-77-1,7732-18-5,7647-01-0 | |
9.18,92.0,1.08 | |
Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
270.29 | |
CHEBI:30808 | |
4L |
10% (w/v) Aqueous Solution | |
8.82,88.0,1.04 | |
Cl3FeH12O6 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
hexahydroxyirontris(ylium) trichloride | |
24380 | |
Poly natural |
Chemical Identifiers
10025-77-1 | |
270.29 | |
Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer | |
CHEBI:30808 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Cl3FeH12O6 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
24380 | |
hexahydroxyirontris(ylium) trichloride |