missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ferric Chloride, 0.025% (w/v) Aqueous Solution, Ricca Chemical
Manufacturer: Ricca Chemical Company RI310016
Specifications
Ferric Chloride | |
10025-77-1,7732-18-5,7647-01-0 | |
0.02,100.0,2.0E-4 | |
Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
270.29 | |
CHEBI:30808 | |
Poly natural | |
Amber |
0.025% (w/v) | |
0.02,98.0,2.0E-4 | |
Cl3FeH12O6 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
hexahydroxyirontris(ylium) trichloride | |
24380 | |
Liquid | |
Aqueous Solution | |
500mL |
Chemical Identifiers
10025-77-1 | |
270.29 | |
Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer | |
CHEBI:30808 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Cl3FeH12O6 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
24380 | |
hexahydroxyirontris(ylium) trichloride |
Promotions
-
Fisherbrand
Spend $250, Get $250 on Ricca Chemical Products
Buy $250 of Ricca Chemical Company products and get $250 more to spend on Ricca items at no additional cost.: 06/30/21